accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH3CPHOONH2

CAS RN 123690-79-9

IUPAC Name: (3-aminopropyl)(diethoxymethyl)phosphinic acid
Synonyms: 3-aminopropyl(diethoxymethyl)phosphinic acid
Molecular Weight (g/mol): 225.23
Molecular Formula: C8H20NO4P
InChi Key: QIIVUOWTHWIXFO-UHFFFAOYNA-N
SMILES: CCOC(OCC)P(O)(=O)CCCN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)