accessibility menu, dialog, popup

UserName

Viewing profile as user:
NFFFFFN

CAS RN 1237431-49-0

IUPAC Name: 6-(1,1,2,2,2-pentafluoroethyl)pyridine-2-carbonitrile
Synonyms: 6-(1,1,2,2,2-pentafluoroethyl)pyridine-2-carbonitrile
Molecular Weight (g/mol): 222.12
Molecular Formula: C8H3F5N2
InChi Key: JHTRASHYGAYPTG-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C1=CC=CC(=N1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results