accessibility menu, dialog, popup

UserName

Viewing profile as user:
NONH2HO

CAS RN 123931-04-4

IUPAC Name: 3-amino-1-hydroxypyrrolidin-2-one
Synonyms: 3-amino-1-hydroxypyrrolidin-2-one
Molecular Weight (g/mol): 116.12
Molecular Formula: C4H8N2O2
InChi Key: HCKUBNLZMKAEIN-UHFFFAOYNA-N
SMILES: NC1CCN(O)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results