CAS RN 124-18-5
n-Decane, 99%
CAS: 124-18-5 Molecular Formula: C10H22 Molecular Weight (g/mol): 142.286 MDL Number: MFCD00008954 InChI Key: DIOQZVSQGTUSAI-UHFFFAOYSA-N Synonym: n-decane,nonane, methyl,decyl hydride,unii-nk85062oiy,ccris 653,hsdb 63,dsstox_cid_4913,decane, analytical standard,dsstox_rid_77577,dsstox_gsid_24913 PubChem CID: 15600 ChEBI: CHEBI:41808 IUPAC Name: decane SMILES: CCCCCCCCCC
Decane, 99+%, pure
CAS: 124-18-5 Molecular Formula: C10H22 Molecular Weight (g/mol): 142.286 MDL Number: MFCD00008954 InChI Key: DIOQZVSQGTUSAI-UHFFFAOYSA-N Synonym: n-decane,nonane, methyl,decyl hydride,unii-nk85062oiy,ccris 653,hsdb 63,dsstox_cid_4913,decane, analytical standard,dsstox_rid_77577,dsstox_gsid_24913 PubChem CID: 15600 ChEBI: CHEBI:41808 IUPAC Name: decane SMILES: CCCCCCCCCC
n-Decane, 99+%
CAS: 124-18-5 Molecular Formula: C10H22 Molecular Weight (g/mol): 142.286 MDL Number: MFCD00008954 InChI Key: DIOQZVSQGTUSAI-UHFFFAOYSA-N Synonym: n-decane,nonane, methyl,decyl hydride,unii-nk85062oiy,ccris 653,hsdb 63,dsstox_cid_4913,decane, analytical standard,dsstox_rid_77577,dsstox_gsid_24913 PubChem CID: 15600 ChEBI: CHEBI:41808 IUPAC Name: decane SMILES: CCCCCCCCCC
Decane 99.0+%, TCI America™
CAS: 124-18-5 Molecular Formula: C10H22 Molecular Weight (g/mol): 142.286 MDL Number: MFCD00008954 InChI Key: DIOQZVSQGTUSAI-UHFFFAOYSA-N Synonym: n-decane,nonane, methyl,decyl hydride,unii-nk85062oiy,ccris 653,hsdb 63,dsstox_cid_4913,decane, analytical standard,dsstox_rid_77577,dsstox_gsid_24913 PubChem CID: 15600 ChEBI: CHEBI:41808 IUPAC Name: decane SMILES: CCCCCCCCCC
Decane, Fisher Chemical
CAS: 124-18-5 Molecular Formula: C10H22 Molecular Weight (g/mol): 142.286 MDL Number: MFCD00008954 InChI Key: DIOQZVSQGTUSAI-UHFFFAOYSA-N Synonym: n-decane,nonane, methyl,decyl hydride,unii-nk85062oiy,ccris 653,hsdb 63,dsstox_cid_4913,decane, analytical standard,dsstox_rid_77577,dsstox_gsid_24913 PubChem CID: 15600 ChEBI: CHEBI:41808 IUPAC Name: decane SMILES: CCCCCCCCCC
Decane, 99%, anhydrous, AcroSeal™
CAS: 124-18-5 Molecular Formula: C10H22 Molecular Weight (g/mol): 142.28 InChI Key: DIOQZVSQGTUSAI-UHFFFAOYSA-N Synonym: n-decane,nonane, methyl,decyl hydride,unii-nk85062oiy,ccris 653,hsdb 63,dsstox_cid_4913,decane, analytical standard,dsstox_rid_77577,dsstox_gsid_24913 PubChem CID: 15600 ChEBI: CHEBI:41808 IUPAC Name: decane SMILES: CCCCCCCCCC
n-Decane, MP Biomedicals™
CAS: 124-18-5 Molecular Formula: C10H22 Molecular Weight (g/mol): 142.29 InChI Key: DIOQZVSQGTUSAI-UHFFFAOYSA-N Synonym: n-decane,nonane, methyl,decyl hydride,unii-nk85062oiy,ccris 653,hsdb 63,dsstox_cid_4913,decane, analytical standard,dsstox_rid_77577,dsstox_gsid_24913 PubChem CID: 15600 ChEBI: CHEBI:41808 IUPAC Name: decane SMILES: CCCCCCCCCC
Decane, BAKER™, J.T. Baker™
CAS: 124-18-5 Molecular Formula: C10H22 Molecular Weight (g/mol): 142.286 InChI Key: DIOQZVSQGTUSAI-UHFFFAOYSA-N Synonym: n-decane,nonane, methyl,decyl hydride,unii-nk85062oiy,ccris 653,hsdb 63,dsstox_cid_4913,decane, analytical standard,dsstox_rid_77577,dsstox_gsid_24913 PubChem CID: 15600 ChEBI: CHEBI:41808 IUPAC Name: decane SMILES: CCCCCCCCCC
Decane, ≥99%, Reagent Grade, Solstice
CAS: 124-18-5 Molecular Formula: C10H22 Molecular Weight (g/mol): 142.286 MDL Number: MFCD00008954 InChI Key: DIOQZVSQGTUSAI-UHFFFAOYSA-N Synonym: n-decane,nonane, methyl,decyl hydride,unii-nk85062oiy,ccris 653,hsdb 63,dsstox_cid_4913,decane, analytical standard,dsstox_rid_77577,dsstox_gsid_24913 PubChem CID: 15600 ChEBI: CHEBI:41808 IUPAC Name: decane SMILES: CCCCCCCCCC
Reference Material for Flash Point Certified by The Japan Petroleum Institute, Decane, TCI America™
CAS: 124-18-5 Molecular Formula: C10H22 Molecular Weight (g/mol): 142.286 MDL Number: MFCD00008954 InChI Key: DIOQZVSQGTUSAI-UHFFFAOYSA-N Synonym: n-decane,nonane, methyl,decyl hydride,unii-nk85062oiy,ccris 653,hsdb 63,dsstox_cid_4913,decane, analytical standard,dsstox_rid_77577,dsstox_gsid_24913 PubChem CID: 15600 ChEBI: CHEBI:41808 IUPAC Name: decane SMILES: CCCCCCCCCC
Decane 99.5+%, TCI America™
CAS: 124-18-5 Molecular Formula: C10H22 Molecular Weight (g/mol): 142.286 MDL Number: MFCD00008954 InChI Key: DIOQZVSQGTUSAI-UHFFFAOYSA-N Synonym: n-decane,nonane, methyl,decyl hydride,unii-nk85062oiy,ccris 653,hsdb 63,dsstox_cid_4913,decane, analytical standard,dsstox_rid_77577,dsstox_gsid_24913 PubChem CID: 15600 ChEBI: CHEBI:41808 IUPAC Name: decane SMILES: CCCCCCCCCC
Decane, SPEX CertiPrep™
CAS: 124-18-5 Molecular Formula: C10H22 Molecular Weight (g/mol): 142.286 InChI Key: DIOQZVSQGTUSAI-UHFFFAOYSA-N PubChem CID: 15600 ChEBI: CHEBI:41808 IUPAC Name: decane SMILES: CCCCCCCCCC
Sigma Aldrich Decane
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
MP Biomedicals, Inc Decamide Capramide, MP Biomedicals
CAS: 124-18-5 Molecular Formula: C10H22 Molecular Weight (g/mol): 142.286 InChI Key: DIOQZVSQGTUSAI-UHFFFAOYSA-N Synonym: n-decane,nonane, methyl,decyl hydride,unii-nk85062oiy,ccris 653,hsdb 63,dsstox_cid_4913,decane, analytical standard,dsstox_rid_77577,dsstox_gsid_24913 PubChem CID: 15600 ChEBI: CHEBI:41808 IUPAC Name: decane SMILES: CCCCCCCCCC
- 1
- 2