Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decane, ≥99%, Reagent Grade, Solstice

Catalog No. 60047980 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-980 500 mL Glass bottle
60-047-979 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-980 Supplier Solstice Supplier No. D901500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-18-5
Molecular Formula C10H22
Molecular Weight (g/mol) 142.286
MDL Number MFCD00008954
InChI Key DIOQZVSQGTUSAI-UHFFFAOYSA-N
Synonym n-decane, nonane, methyl, decyl hydride, unii-nk85062oiy, ccris 653, hsdb 63, dsstox_cid_4913, decane, analytical standard, dsstox_rid_77577, dsstox_gsid_24913
PubChem CID 15600
ChEBI CHEBI:41808
IUPAC Name decane
SMILES CCCCCCCCCC

Specifications

Boiling Range 174° - 180°C
Melting Point -27°C
Color Colorless
Density 0.73g/mL at 25°C
Explosion Limit 0.026
Flash Point 46°C (114.8°F)
Ignition Point 410°F
Odor Odorless
Packaging Glass bottle
Linear Formula CH3(CH2)8CH3
Quantity 500 mL
UN Number UN2247
Beilstein 1696981
Vapor Density 4.9 (vs air)
Vapor Pressure 3.77mmHg (37.7°C), 1mmHg (16.5°C)
Formula Weight 142.28g/mol
Percent Purity ≥99%
Chemical Name or Material Decane
Show More Show Less
Hazard Statement P301 + P310-P331
DOTInformation 3/PGIII
RTECSNumber 204-686-4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.