accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 124344-94-1

IUPAC Name: 1-methyl-5-phenyl-1H-pyrazole-3-carbaldehyde
Synonyms: 1-methyl-5-phenylpyrazole-3-carbaldehyde
Molecular Weight (g/mol): 186.21
Molecular Formula: C11H10N2O
InChi Key: AGEKFZHCMVDIFA-UHFFFAOYSA-N
SMILES: CN1N=C(C=O)C=C1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results