accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOHNNFNHOHCl

CAS RN 1244640-48-9

IUPAC Name: N-{2-[1-(3-fluorophenyl)-4-oxo-1,3,8-triazaspiro[4.5]decan-8-yl]ethyl}naphthalene-2-carboxamide hydrochloride
Synonyms: N-{2-[1-(3-fluorophenyl)-4-oxo-1,3,8-triazaspiro[4.5]decan-8-yl]ethyl}naphthalene-2-carboxamide hydrochloride
Molecular Weight (g/mol): 482.98
Molecular Formula: C26H28ClFN4O2
InChi Key: RYLAMDMOILNBKN-UHFFFAOYSA-N
SMILES: Cl.FC1=CC=CC(=C1)N1CNC(=O)C11CCN(CCNC(=O)C2=CC=C3C=CC=CC3=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)