accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHFFFFFFOHFFF

CAS RN 1246525-60-9

IUPAC Name: N-[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]-4-(trifluoromethyl)benzamide
Synonyms: N-[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]-4-(trifluoromethyl)benzamide
Molecular Weight (g/mol): 431.26
Molecular Formula: C17H10F9NO2
InChi Key: DUXWIYXHHGNUJU-UHFFFAOYSA-N
SMILES: OC(C1=CC=C(NC(=O)C2=CC=C(C=C2)C(F)(F)F)C=C1)(C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results