accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSSFFNOO

CAS RN 1247819-59-5

IUPAC Name: 1-{5-[(2,4-difluorophenyl)sulfanyl]-4-nitrothiophen-2-yl}ethan-1-one
Synonyms: 1-{5-[(2,4-difluorophenyl)sulfanyl]-4-nitrothiophen-2-yl}ethanone
Molecular Weight (g/mol): 315.31
Molecular Formula: C12H7F2NO3S2
InChi Key: RMAMGGNACJHXHO-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(=C(SC2=C(F)C=C(F)C=C2)S1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results