accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2NNH

CAS RN 1250405-19-6

IUPAC Name: (3-amino-2-methylpropyl)(benzyl)amine
Synonyms: (3-amino-2-methylpropyl)(benzyl)amine
Molecular Weight (g/mol): 178.28
Molecular Formula: C11H18N2
InChi Key: RICFCTFKNQIVMT-UHFFFAOYNA-N
SMILES: CC(CN)CNCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results