Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyl-2-methyl-1,3-propanediamine 97.0+%, TCI America™
SDP

Catalog No. B26875G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B26875G Supplier TCI America Supplier No. B26875G

Chemical Identifiers

CAS 1250405-19-6
Molecular Formula C11H18N2
Molecular Weight (g/mol) 178.279
InChI Key RICFCTFKNQIVMT-UHFFFAOYSA-N
Synonym N-(3-Amino-2-methylpropyl)benzylamine, 3-Benzylamino-2-methylpropylamine
PubChem CID 44629768
IUPAC Name N'-benzyl-2-methylpropane-1,3-diamine
SMILES CC(CN)CNCC1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 178.28
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Benzyl-2-methyl-1,3-propanediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.