accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNNN

CAS RN 1251503-35-1

IUPAC Name: 2-(2-{5H-pyrido[3,2-b]indol-8-yl}phenyl)pyridine
Synonyms: 2-(2-{5H-pyrido[3,2-b]indol-8-yl}phenyl)pyridine
Molecular Weight (g/mol): 321.38
Molecular Formula: C22H15N3
InChi Key: ROQWGQBJRGUPAG-UHFFFAOYSA-N
SMILES: N1C2=C(C=C(C=C2)C2=CC=CC=C2C2=CC=CC=N2)C2=C1C=CC=N2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results