CAS RN 125313-65-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Ro-31-7549, Monohydrate
A cell permeable, reversible, selective protein kinase C (PKC) inhibitor (IC50 = 158 nM for rat brain PKC) that acts at the ATP binding site of PKC.