CAS RN 125316-60-1
IUPAC Name:
6-[3-(adamantan-1-yl)-4-hydroxyphenyl]naphthalene-2-carboxylic acid
Synonyms:
6-[3-(adamantan-1-yl)-4-hydroxyphenyl]naphthalene-2-carboxylic acid
Molecular Weight (g/mol):
398.5
Molecular Formula:
C27H26O3
InChi Key:
LDGIHZJOIQSHPB-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=C2C=C(C=CC2=C1)C1=CC=C(O)C(=C1)C12CC3CC(CC(C3)C1)C2
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ Apoptosis Activator VI, CD437/AHPN
The Apoptosis Activator VI, CD437/AHPN, also referenced under CAS 125316-60-1, modulates Apoptosis. This small molecule/inhibitor is primarily used for Cancer applications.
2
R&D Systems™ CD 437
CAS: 125316-60-1 Molecular Formula: C27H26O3