accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNCH3NNNNNNClClRu

CAS RN 1256362-30-7

IUPAC Name: λ²-ruthenium(2+) bis(2,2'-bipyridine) bis(3-(1-methylpyrrolidin-2-yl)pyridine) dichloride
Synonyms: λ²-ruthenium(2+) bis(bipyridyl) bis(nicotine) dichloride
Molecular Weight (g/mol): 808.82
Molecular Formula: C40H44Cl2N8Ru
InChi Key: BQXVYFYPLMVART-UHFFFAOYNA-L
SMILES: [Cl-].[Cl-].[Ru++].CN1CCCC1C1=CN=CC=C1.CN1CCCC1C1=CN=CC=C1.C1=CC=C(N=C1)C1=CC=CC=N1.C1=CC=C(N=C1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results