accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNNNONOOCl

CAS RN 1257256-44-2

IUPAC Name: N-(3-chloro-4-methylphenyl)-5-(4-nitrophenyl)-1,3,4-oxadiazol-2-amine
Synonyms: N-(3-chloro-4-methylphenyl)-5-(4-nitrophenyl)-1,3,4-oxadiazol-2-amine
Molecular Weight (g/mol): 330.73
Molecular Formula: C15H11ClN4O3
InChi Key: JYCNWQGNEJYDQS-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C=C(NC2=NN=C(O2)C2=CC=C(C=C2)[N+]([O-])=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)