accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3OSiOSiOSiOSiOOSiOSiOOSiOOSiOOOSiCH3H3COSiH3CCH3OSiCH3CH3OSiH3CH3COSiH3CCH3OSiCH3CH3OSiH3CH3C

CAS RN 125756-69-6

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 1009.9
Molecular Formula: C16H48O20Si16
InChi Key: NKAADCIRFJPCOM-UHFFFAOYSA-N
SMILES: C[Si](C)O[Si]12O[Si]3(O[Si](C)C)O[Si]4(O[Si](C)C)O[Si](O[Si](C)C)(O1)O[Si]1(O[Si](C)C)O[Si](O[Si](C)C)(O2)O[Si](O[Si](C)C)(O3)O[Si](O[Si](C)C)(O4)O1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)