accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)CH3(S)NHOHNO(S)H2NONHO(S)NH2HNO(S)NHO(S)HOHNO(S)NO(R)HSNHOHNO(R)HSNHOHNO(S)NHNH2NHHNO(S)OHOHNO(S)NHO(S)HNNH2NHNHO(S)HONHOHNO(S)HNNH2HNNHO(S)NHNHNO(S)OHONHO(S)HNNH2NHHNO(S)HNHNO(S)HNNH2NHNHO(S)NH2HNO(S)H2NNHO(S)OHHNO(R)HSNHO(S)H2NOHNO(S)NHO(S)OHOHNONHOHNO(S)HONHO(R)SHHNONHO(S)HNO(S)NHO(S)HNO(S)NO(S)NHO(S)NH2OHNO(S)OHHNO(S)OHNHO(S)NH2OHNO(S)NH2ONHO(S)H2NOHNO(S)HNNNHO(R)SHHNO(S)OHONHO(S)NH2CH3CH3(R)H3COH(R)CH3OH(R)CH3HO(R)CH3OH(R)CH3HO(R)CH3HOCH3H3COHN(S)OHOONH(S)(S)CH3H3COHN(S)NH2OONH(R)HSOHN(R)HSONH(S)(R)H3COHOHN(S)(R)H3COHONH(S)HOOOHN(S)HNNH2HNONH(R)HSOHN(S)NH2OONH(S)H2NOHOO

CAS RN 12584-83-7

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 6956.65
Molecular Formula: C277H443N97O98S8
InChi Key: BVGLZNQZEYAYBJ-QWZQWHGGSA-N
SMILES: CC[C@H](C)[C@H](NC(=O)CNC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CCCCN)NC(=O)[C@@H](NC(=O)[C@H](CO)NC(=O)[C@@H]1CCCN1C(=O)[C@H](CS)NC(=O)CNC(=O)[C@H](CS)NC(=O)CNC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)CNC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC1=CN=CN1)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)[C@H](CS)NC(=O)[C@H](CC(N)=O)NC(=O)[C@@H](NC(=O)[C@H](CCC(O)=O)NC(=O)CNC(=O)CNC(=O)[C@H](CO)NC(=O)[C@H](CS)NC(=O)CNC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H]1CCCN1C(=O)[C@@H](NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CO)NC(=O)[C@H](CO)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CC1=CN=CN1)NC(=O)[C@H](CS)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@@H](N)CC(C)C)[C@@H](C)O)[C@@H](C)O)[C@@H](C)O)[C@@H](C)O)[C@@H](C)O)[C@@H](C)O)C(C)C)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CS)C(=O)N[C@@H](CS)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CS)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CC(N)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results