accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 126251-92-1

IUPAC Name: [2'-(diphenylphosphanyl)-[1,1'-binaphthalen]-2-yl]diphenylphosphane benzene chlororutheniumylium chloride
Synonyms: benzene binap chlororutheniumylium chloride
Molecular Weight (g/mol): 872.77
Molecular Formula: C50H38Cl2P2Ru
InChi Key: ZXBWEYDPYJZEFT-UHFFFAOYSA-L
SMILES: [Cl-].Cl[Ru+].C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results