accessibility menu, dialog, popup

UserName

Viewing profile as user:
HCOOOOHNOSNHOHN

CAS RN 1262681-31-1

IUPAC Name: 5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}-N-(3,6,9,12-tetraoxapentadec-14-yn-1-yl)pentanamide
Synonyms: 5-{2-oxo-hexahydrothieno[3,4-d]imidazol-4-yl}-N-(3,6,9,12-tetraoxapentadec-14-yn-1-yl)pentanamide
Molecular Weight (g/mol): 457.59
Molecular Formula: C21H35N3O6S
InChi Key: SKMJWNZZFUDLKQ-UHFFFAOYNA-N
SMILES: O=C(CCCCC1SCC2NC(=O)NC12)NCCOCCOCCOCCOCC#C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results