accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NBrNH2

CAS RN 126325-47-1

IUPAC Name: 6-bromo-2-methylpyridin-3-amine
Synonyms: 6-bromo-2-methylpyridin-3-amine
Molecular Weight (g/mol): 187.04
Molecular Formula: C6H7BrN2
InChi Key: UBTQJSZELCWGCN-UHFFFAOYSA-N
SMILES: CC1=C(N)C=CC(Br)=N1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)