accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOH3CBr

CAS RN 126717-59-7

IUPAC Name: 3-bromo-6-methoxy-2-methylpyridine
Synonyms: 3-bromo-6-methoxy-2-methylpyridine
Molecular Weight (g/mol): 202.05
Molecular Formula: C7H8BrNO
InChi Key: VWNXCCTWVAQAPL-UHFFFAOYSA-N
SMILES: COC1=NC(C)=C(Br)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)