Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-6-methoxy-2-methylpyridine, 98%, Thermo Scientific™

Catalog No. AAH6196203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6196203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6196203 Supplier Thermo Scientific Chemicals Supplier No. H6196203

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126717-59-7
Molecular Formula C7H8BrNO
Molecular Weight (g/mol) 202.051
MDL Number MFCD03094943
InChI Key VWNXCCTWVAQAPL-UHFFFAOYSA-N
Synonym 5-bromo-2-methoxy-6-picoline, 3-bromo-6-methoxy-2-picoline, 3-bromo-6-methoxy-2-methyl-pyridine, 2-methoxy-5-bromo-6-picoline, 2-methyl-3-bromo-6-methoxypyridine, 5-bromo-2-methoxy-6-methyl pyridine, 2-methoxy-5-bromo-6-methylpyridine, 5-bromo-2-methoxy-6-methylpyridine, 5-bromo-6-methyl-2-methoxypyridine, pyridine, 3-bromo-6-methoxy-2-methyl
PubChem CID 10798146
IUPAC Name 3-bromo-6-methoxy-2-methylpyridine
SMILES CC1=C(C=CC(=N1)OC)Br

Specifications

Color Colorless
Quantity 1 g
Formula Weight 202.05
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Bromo-6-methoxy-2-methylpyridine
Hazard Category H302-H318
Hazard Statement GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.