accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 126726-62-3

IUPAC Name: 4,4,5,5-tetramethyl-2-(prop-1-en-2-yl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(prop-1-en-2-yl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 168.04
Molecular Formula: C9H17BO2
InChi Key: SVSUYEJKNSMKKW-UHFFFAOYSA-N
SMILES: CC(=C)B1OC(C)(C)C(C)(C)O1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)