Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isopropenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 90%

Catalog No. AC430450050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC430450050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC430450050 Supplier Thermo Scientific Chemicals Supplier No. 430450050
  • $163.40
    $138.65 / Each of 1

    Save $24.75

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 126726-62-3 | C9H17BO2 | 168.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126726-62-3
Molecular Formula C9H17BO2
Molecular Weight (g/mol) 168.04
MDL Number MFCD08276843
InChI Key SVSUYEJKNSMKKW-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-prop-1-en-2-yl-1,3,2-dioxaborolane, isopropenylboronic acid pinacol ester, 2-isopropenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-isopropenylboronic acid pinacol ester, 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl isopropene, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-methylethenyl, 4,4,5,5-tetramethyl-2-1-methylethenyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-isopropenyl-1,3,2-dioxaborolane, isopropenylboronic acid pinacol ester, stabilized with phenothiazine, pubchem17392
PubChem CID 10997426
IUPAC Name 4,4,5,5-tetramethyl-2-prop-1-en-2-yl-1,3,2-dioxaborolane
SMILES CC(=C)B1OC(C)(C)C(C)(C)O1

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Brown to Colorless
Density 0.8800g/mL
Boiling Point 47.0°C (9.0 Mbar)
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 0.88
Stabilizer 1% phenothiazine
Formula Weight 168.04
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 2-Isopropenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.