CAS RN 126766-32-3
IUPAC Name:
(2E)-but-2-enedioic acid; methyl 4-[2-(3,4-dichlorophenyl)acetyl]-3-[(pyrrolidin-1-yl)methyl]piperazine-1-carboxylate
Synonyms:
fumaric acid; methyl 4-[2-(3,4-dichlorophenyl)acetyl]-3-(pyrrolidin-1-ylmethyl)piperazine-1-carboxylate
Molecular Weight (g/mol):
530.4
Molecular Formula:
C23H29Cl2N3O7
InChi Key:
ABTNETSDXZBJTE-WLHGVMLRNA-N
SMILES:
OC(=O)\C=C\C(O)=O.COC(=O)N1CCN(C(CN2CCCC2)C1)C(=O)CC1=CC=C(Cl)C(Cl)=C1
1
–
1
of
1
results
1
R&D Systems™ GR 89696 fumarate
CAS: 126766-32-3 Molecular Formula: C23H29Cl2N3O7