accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1271-24-5

IUPAC Name: Bis(cyclopentadienyl)chromium
Synonyms: chromocene
Molecular Weight (g/mol): 182.19
Molecular Formula: C10H10Cr
InChi Key: TYYBBNOTQFVVKN-UHFFFAOYSA-N
SMILES: [Cr++].[CH-]1C=CC=C1.[CH-]1C=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results