accessibility menu, dialog, popup

UserName

Viewing profile as user:
FeCH

CAS RN 1271-47-2

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 210.06
Molecular Formula: C12H10Fe
InChi Key:
SMILES: [Fe].c1cccc1.C#Cc1cccc1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)