accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1271-51-8

FeCH2

CAS RN 1271-51-8

IUPAC Name: Vinylferrocene
Synonyms:
Molecular Weight (g/mol): 212.07
Molecular Formula: C12H12Fe
InChi Key:
SMILES: [Fe].c1cccc1.C=Cc1cccc1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (3)
  • (2)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (1)
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)