Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinylferrocene 97.0+%, TCI America™
SDP

Catalog No. V00641G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V00641G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V00641G Supplier TCI America Supplier No. V00641G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1271-51-8
Molecular Formula C12H12Fe
Molecular Weight (g/mol) 212.07
MDL Number MFCD00001434
InChI Key VGBAECKRTWHKHC-UHFFFAOYSA-N
IUPAC Name Vinylferrocene
SMILES [Fe].c1cccc1.C=Cc1cccc1

Specifications

Melting Point 53°C
Quantity 1 g
UN Number 1325
Formula Weight 212.07
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Vinylferrocene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.