accessibility menu, dialog, popup

UserName

Viewing profile as user:
FeOCH3

CAS RN 1271-55-2

IUPAC Name: 1-Acetylferrocene
Synonyms:
Molecular Weight (g/mol): 228.07
Molecular Formula: C12H12FeO
InChi Key:
SMILES: [Fe].c1cccc1.CC(=O)c1cccc1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results