accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrHNNHSOON(E)

CAS RN 127243-85-0

IUPAC Name: N-(2-{[(2E)-3-(4-bromophenyl)prop-2-en-1-yl]amino}ethyl)isoquinoline-5-sulfonamide
Synonyms: N-(2-{[(2E)-3-(4-bromophenyl)prop-2-en-1-yl]amino}ethyl)isoquinoline-5-sulfonamide
Molecular Weight (g/mol): 446.36
Molecular Formula: C20H20BrN3O2S
InChi Key: ZKZXNDJNWUTGDK-NSCUHMNNSA-N
SMILES: BrC1=CC=C(\C=C\CNCCNS(=O)(=O)C2=C3C=CN=CC3=CC=C2)C=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results