accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOHONNN

CAS RN 1272755-25-5

IUPAC Name: 5-azido-4-(benzyloxy)-2-[(benzyloxy)methyl]-6-(4-methoxyphenoxy)oxan-3-ol
Synonyms: 5-azido-4-(benzyloxy)-2-[(benzyloxy)methyl]-6-(4-methoxyphenoxy)oxan-3-ol
Molecular Weight (g/mol): 491.54
Molecular Formula: C27H29N3O6
InChi Key: ZTAXYCGZXSYIMV-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC(COCC3=CC=CC=C3)C(O)C(OCC3=CC=CC=C3)C2N=[N+]=[N-])C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results