CAS RN 127299-93-8
CAS RN 127299-93-8
IUPAC Name:
sodium 2-[4-(2-{[2-(3-chlorophenyl)-2-hydroxyethyl]amino}propyl)phenoxy]acetate
Synonyms:
sodium 2-[4-(2-{[2-(3-chlorophenyl)-2-hydroxyethyl]amino}propyl)phenoxy]acetate
Molecular Weight (g/mol):
385.82
Molecular Formula:
C19H21ClNNaO4
InChi Key:
SNJIJYKMYQRHRC-UHFFFAOYNA-M
SMILES:
[Na+].CC(CC1=CC=C(OCC([O-])=O)C=C1)NCC(O)C1=CC(Cl)=CC=C1
1
–
1
of
1
results
1
Tocris Bioscience™ BRL 37344, sodium salt
CAS: 127299-93-8 Molecular Formula: C19H21ClNNaO4