accessibility menu, dialog, popup

UserName

Viewing profile as user:
(S)O(R)(S)(S)(R)(S)(S)(R)(R)OOHHOOOHHOHOOHHOOHOHOmonmon

CAS RN 12738-64-6

IUPAC Name: (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol; benzoic acid
Synonyms: benzoic acid; sucrose
Molecular Weight (g/mol): 464.42
Molecular Formula: C19H28O13
InChi Key: GYUJITDJMFVEPZ-CKQFFSBTNA-N
SMILES: OC(=O)C1=CC=CC=C1.OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)