Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sucrose octabenzoate, 98%, Thermo Scientific™

Catalog No. AAJ6608322
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6608322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6608322 Supplier Thermo Scientific Chemicals Supplier No. J6608322

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12738-64-6
Molecular Formula C19H28O13
Molecular Weight (g/mol) 464.42
MDL Number MFCD00047542
InChI Key GYUJITDJMFVEPZ-CKQFFSBTNA-N
Synonym Sucrose benzoate

Specifications

Melting Point 93°C to 100°C
Color White
Quantity 100 g
Formula Weight 1175.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Sucrose octabenzoate
EINECSNumber 235-795-5
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.