Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sucrose Benzoate, MP Biomedicals

Catalog No. ICN218913 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN218913 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN218913 Supplier MP Biomedicals, Inc Supplier No. 0521891380

Chemical Identifiers

CAS 12738-64-6
Molecular Formula C19H28O13
Molecular Weight (g/mol) 464.42
InChI Key GYUJITDJMFVEPZ-CKQFFSBTNA-N

Specifications

Quantity 100 g
Chemical Name or Material Sucrose Benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.