accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOClHOOHOO(E)(E)

CAS RN 12772-57-5

IUPAC Name: (9E,11E)-16-chloro-17,19-dihydroxy-4-methyl-3,7-dioxatricyclo[13.4.0.0⁶,⁸]nonadeca-1(19),9,11,15,17-pentaene-2,13-dione
Synonyms: radicicol
Molecular Weight (g/mol): 364.78
Molecular Formula: C18H17ClO6
InChi Key: WYZWZEOGROVVHK-ZUVMSYQZNA-N
SMILES: CC1CC2OC2\C=C\C=C\C(=O)CC2=C(Cl)C(O)=CC(O)=C2C(=O)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)