Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Radicicol
SDP

Catalog No. 15891 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-891 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 15-891 Supplier R&D Systems™ Supplier No. 1589/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Serine protease inhibitor; inhibits tryptase

Antifungal antibiotic. Inhibits heat shock protein 90 (Hsp90) activity by binding to the ATP-binding pocket, subsequently suppressing cell transformation induced by src, ras and mos. Represses estrogen receptor transcriptional activity and inhibits angiogenesis.

Chemical Identifiers

CAS 12772-57-5
Molecular Formula C18H17ClO6

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 364.78
Purity >98%
Molecular Weight (g/mol) 364.778
Chemical Name or Material Radicicol
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.