accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONHOSFCl

CAS RN 1279713-77-7

IUPAC Name: 2-[(3-chloro-6-fluoro-1-benzothiophen-2-yl)formamido]-3-phenylpropanoic acid
Synonyms: 2-[(3-chloro-6-fluoro-1-benzothiophen-2-yl)formamido]-3-phenylpropanoic acid
Molecular Weight (g/mol): 377.81
Molecular Formula: C18H13ClFNO3S
InChi Key: IAASQMCXDRISAV-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC=CC=C1)NC(=O)C1=C(Cl)C2=CC=C(F)C=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)