CAS RN 1285515-21-0
IUPAC Name:
2-(benzyloxy)-5-(2-fluoropyridin-4-yl)-N-(pyridin-3-yl)benzamide
Synonyms:
2-(benzyloxy)-5-(2-fluoropyridin-4-yl)-N-(pyridin-3-yl)benzamide
Molecular Weight (g/mol):
399.43
Molecular Formula:
C24H18FN3O2
InChi Key:
WCIGMFCFPXZRMQ-UHFFFAOYSA-N
SMILES:
FC1=NC=CC(=C1)C1=CC(C(=O)NC2=CN=CC=C2)=C(OCC2=CC=CC=C2)C=C1
1
–
1
of
1
results
1
Tocris Bioscience™ GSK2578215A
Potent, selective LRRK2 inhibitor; brain penetrant