accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNFNOO

CAS RN 1285896-76-5

IUPAC Name: 1-ethyl-4-(4-fluoro-2-nitrophenyl)piperazine
Synonyms: 1-ethyl-4-(4-fluoro-2-nitrophenyl)piperazine
Molecular Weight (g/mol): 253.28
Molecular Formula: C12H16FN3O2
InChi Key: IWKXGRRMWASTNC-UHFFFAOYSA-N
SMILES: CCN1CCN(CC1)C1=C(C=C(F)C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results