accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 129-16-8

IUPAC Name: merbromin
Synonyms: merbromin
Molecular Weight (g/mol): 751.66
Molecular Formula: C20H9Br2HgNa2O6
InChi Key: BFMYDTVEBKDAKJ-UHFFFAOYNA-L
SMILES: O.[Na+].[Na+].[O-]C1=C(Br)C=C2C(OC3=C(C=C(Br)C([O-])=C3[Hg])C22OC(=O)C3=CC=CC=C23)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)