Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercurochrome, 24-27% Mercury

Catalog No. AC190111000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC190111000 100 g Glass bottle
AC190110250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. AC190111000 Supplier Thermo Scientific Chemicals Supplier No. 190111000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

CAS: 129-16-8 | C20H8Br2HgNa2O6 | 750.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129-16-8
Molecular Formula C20H8Br2HgNa2O6
Molecular Weight (g/mol) 750.65
MDL Number MFCD00013081
InChI Key QJJYLIJNUFJILQ-UHFFFAOYSA-N
Synonym Merbromin, 2, 7-Dibromo-4-hydroxymercurifluoresceine disodium salt
PubChem CID 54598522
IUPAC Name sodium;[2,7-dibromo-9-(2-carboxyphenyl)-3-hydroxy-6-oxoxanthen-4-yl]mercury;hydrate
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C=C3OC4=C(C(=C(C=C24)Br)O)[Hg])Br)C(=O)O.O.[Na+]

Specifications

Melting Point >300°C
Color Green
Packaging Glass bottle
Quantity 100 g
Merck Index 15, 5934
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol,acetone,ether
Formula Weight 750.65
Percent Purity 24 to 27% (Hg), 18 to 22.5% (Br2)
Physical Form Glimmering Scales, Granules or Crystals
Chemical Name or Material Mercurochrome
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Fatal in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-933-6
RTECSNumber LM5250000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.