accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NHOCl

CAS RN 1293014-05-7

IUPAC Name: 4-(chloromethyl)-N-(2-ethylhexyl)benzamide
Synonyms: 4-(chloromethyl)-N-(2-ethylhexyl)benzamide
Molecular Weight (g/mol): 281.82
Molecular Formula: C16H24ClNO
InChi Key: JLQFBEBBEBVSIV-UHFFFAOYNA-N
SMILES: CCCCC(CC)CNC(=O)C1=CC=C(CCl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results