accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 129541-41-9

IUPAC Name: sodium 6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate
Synonyms: sodium 6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate
Molecular Weight (g/mol): 444.59
Molecular Formula: C14H12BrClNNaO7
InChi Key: IBLSVGDGSKUDCT-UHFFFAOYNA-M
SMILES: [Na+].OC1C(O)C(OC2=CNC3=CC=C(Br)C(Cl)=C23)OC(C1O)C([O-])=O
Physical Form 
  • (2)
  • (6)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (6)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)