Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-4-Chloro-3-Indoyl-β-D-Glucuronide sodium salt, 98%, Molecular biology reagent grade, For TLC analysis, MP Biomedicals™

Catalog No. ICN19481380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19481380 100 mg
ICN19481310 10 mg
ICN19481325 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19481380 Supplier MP Biomedicals, Inc Supplier No. 0219481380

missing translation for 'validMainframeMsg'

5-Bromo-4-chloro-3-indolyl-β-D-glucuronide sodium salt is β-glucuronidase substrate which forms an intense blue precipitate upon enzymatic action

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129541-41-9
Molecular Formula C14H12BrClNNaO7
Molecular Weight (g/mol) 444.594
MDL Number MFCD00135782
InChI Key IBLSVGDGSKUDCT-ILIJQVQCSA-M
Synonym x-gluc sodium salt, x-glca sodium salt, 5-bromo-4-chloro-3-indolyl beta-d-glucopyranosiduronic acid sodium salt
PubChem CID 53384407
IUPAC Name sodium;(2R,3S,4S,5S,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate
SMILES C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)C(=O)[O-])O)O)O)Cl)Br.[Na+]

Specifications

Color White
Quantity 100 mg
Formula Weight 444.6
Grade Molecular Biology Reagent
Physical Form Powder
Chemical Name or Material 5-Bromo-4-Chloro-3-Indoyl-β-D-Glucuronide sodium salt, For TLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.