accessibility menu, dialog, popup

UserName

Viewing profile as user:
NSNFBrBrF

CAS RN 1295502-53-2

IUPAC Name: 4,7-dibromo-5,6-difluoro-2,1,3-benzothiadiazole
Synonyms: 4,7-dibromo-5,6-difluoro-2,1,3-benzothiadiazole
Molecular Weight (g/mol): 329.94
Molecular Formula: C6Br2F2N2S
InChi Key: HFUBKQHDPJZQIW-UHFFFAOYSA-N
SMILES: FC1=C(Br)C2=NSN=C2C(Br)=C1F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results