accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 129722-12-9

IUPAC Name: 7-{4-[4-(2,3-dichlorophenyl)piperazin-1-yl]butoxy}-1,2,3,4-tetrahydroquinolin-2-one
Synonyms: aripiprazole abilitat aripiprazolum abilify aripiprazol 阿立哌唑
Molecular Weight (g/mol): 448.39
Molecular Formula: C23H27Cl2N3O2
InChi Key: CEUORZQYGODEFX-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(N2CCN(CCCCOC3=CC=C4CCC(=O)NC4=C3)CC2)=C1Cl
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)