Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Aripiprazole
GREENER_CHOICESDP

Catalog No. 558450 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
558450 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Catalog No. 558450 Supplier R&D Systems™ Supplier No. 5584/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'

A Tocris product, part of R&D Systems. High affinity D2 and 5-HT1A receptor partial agonist; also 5-HT2A antagonist

Aripiprazole is a high affinity dopamine D2 and 5-HT1A receptor partial agonist (Ki values are ∽1.64 and 5.59 nM respectively). Also 5-HT2A antagonist (Ki = 8.7 nM). Suppresses morphine-induced emesis in vivo. Atypical antipsychotic.

Specifications

Chemical Name or Material Aripiprazole
CAS 129722-12-9
Synonym 7-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy]-3,4-dihydro-2(1H)-quinolinone
Quantity 50 mg
Solubility Soluble to 100mM in DMSO
Product Line Tocris
Molecular Formula C23H27Cl2N3O2
Formula Weight 448.39
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >99%
Show More Show Less

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.